Thermo Scientific Chemicals

Benzyl bromide, 99%, Thermo Scientific Chemicals

Catalog number: A13535.0B
1000 g, Each
Thermo Scientific Chemicals

Benzyl bromide, 99%, Thermo Scientific Chemicals

Catalog number: A13535.0B
1000 g, Each
Quantity
Catalog number: A13535.0B
also known as A13535-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-39-0
IUPAC Name
(bromomethyl)benzene
Molecular Formula
C7H7Br
InChI Key
AGEZXYOZHKGVCM-UHFFFAOYSA-N
SMILES
BrCC1=CC=CC=C1
Assay (GC)
≥98.5%
Refractive Index
1.5740-1.5770 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless to yellow
Identification (FTIR)
Conforms

Description

Benzyl bromide is widely useful in organic synthesis as a benzyl protecting group for alcohols and carboxylic acids. It is used in the preparation of foaming and frothing agents, quaternary ammonium compounds, peniclins, photographic chemicals, synthetic resins, dyes and plasticizers and esters. It is also involved in the synthesis of (E)-(2-Iodovinyl)benzene by reacting it with diiodomethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl bromide is widely useful in organic synthesis as a benzyl protecting group for alcohols and carboxylic acids. It is used in the preparation of foaming and frothing agents, quaternary ammonium compounds, peniclins, photographic chemicals, synthetic resins, dyes and plasticizers and esters. It is also involved in the synthesis of (E)-(2-Iodovinyl)benzene by reacting it with diiodomethane.

Solubility
Miscible with benzene, carbon tetrachloride, ethanol and ether.

Notes
Light and moisture sensitive. Incompatible with bases, alcohols, amines, oxidizing agents and corrodes steel.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text