4'-Nitroacetanilide, 98%, Thermo Scientific Chemicals
4'-Nitroacetanilide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4'-Nitroacetanilide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A13539.06
also known as A13539-06
Price (USD)/ Each
28.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
28.90
Add to cart
4'-Nitroacetanilide, 98%, Thermo Scientific Chemicals
Catalog numberA13539.06
Price (USD)/ Each
28.90
-
Add to cart
Chemical Identifiers
CAS104-04-1
IUPAC NameN-(4-nitrophenyl)acetamide
Molecular FormulaC8H8N2O3
InChI KeyNQRLPDFELNCFHW-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)212.0-218.0?C
Appearance (Color)Yellow to green to brown
FormPowder or lumps
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only