Thermo Scientific Chemicals

2,4-Dihydroxybenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: A13545.36
500 g, Each
Thermo Scientific Chemicals

2,4-Dihydroxybenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: A13545.36
500 g, Each
Quantity
Catalog number: A13545.36
also known as A13545-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
89-86-1
IUPAC Name
2,4-dihydroxybenzoic acid
Molecular Formula
C7H6O4
InChI Key
UIAFKZKHHVMJGS-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(O)C=C1O
Appearance (Color)
White to cream
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (Silylated GC)
≥96.0%
Form
Powder

Description

2,4-Dihydroxybenzoic acid is used in Intermediates, Plasticizers, Plastic and Rubber Products, a organic building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dihydroxybenzoic acid is used in Intermediates, Plasticizers, Plastic and Rubber Products, a organic building block.

Solubility
Soluble in water(70g/L).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text