Thermo Scientific Chemicals

3,4-Dimethoxyaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A13548.30
250 g, Each
Thermo Scientific Chemicals

3,4-Dimethoxyaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A13548.30
250 g, Each
Quantity
Catalog number: A13548.30
also known as A13548-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6315-89-5
IUPAC Name
3,4-dimethoxyaniline
Molecular Formula
C8H11NO2
InChI Key
LGDHZCLREKIGKJ-UHFFFAOYSA-N
SMILES
COC1=CC=C(N)C=C1OC
Melting Point (clear melt)
84.5-90.5?C
Appearance (Color)
Cream to brown
Assay (GC)
≥98.0%
Form
Powder

Description

3,4-Dimethoxyaniline is commonly used as fine chemicals, used for pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethoxyaniline is commonly used as fine chemicals, used for pharmaceutical intermediates.

Solubility
Slightly soluble in water

Notes
Air Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text