4-Chloro-3-nitrobenzaldehyde, 97%, Thermo Scientific Chemicals
4-Chloro-3-nitrobenzaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chloro-3-nitrobenzaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number A13549.18
also known as A13549-18
Price (USD)/ Each
168.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
168.00
Add to cart
4-Chloro-3-nitrobenzaldehyde, 97%, Thermo Scientific Chemicals
Catalog numberA13549.18
Price (USD)/ Each
168.00
-
Add to cart
Chemical Identifiers
CAS16588-34-4
IUPAC Name4-chloro-3-nitrobenzaldehyde
Molecular FormulaC7H4ClNO3
InChI KeyHETBKLHJEWXWBM-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(C=O)=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream or pale yellow
Assay (GC)≥96.0%
Melting Point (clear melt)56.0-65.0?C
FormPowder or lumps
Free acid (titration)≤1.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only