2-(Methoxycarbonyl)benzenesulfonamide, 98%, Thermo Scientific Chemicals
2-(Methoxycarbonyl)benzenesulfonamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Methoxycarbonyl)benzenesulfonamide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number A13553.22
also known as A13553-22
Price (USD)/ Each
73.65
Online exclusive
81.30 
Save 7.65 (9%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
73.65
Online exclusive
81.30 
Save 7.65 (9%)
Add to cart
2-(Methoxycarbonyl)benzenesulfonamide, 98%, Thermo Scientific Chemicals
Catalog numberA13553.22
Price (USD)/ Each
73.65
Online exclusive
81.30 
Save 7.65 (9%)
-
Add to cart
Chemical Identifiers
CAS57683-71-3
IUPAC Namemethyl 2-sulfamoylbenzoate
Molecular FormulaC8H9NO4S
InChI KeyVSOOBQALJVLTBH-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC=C1S(N)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms
Melting Point123.0-129.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only