Thermo Scientific Chemicals

2-Iodobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A13558.18
50 g, Each
Thermo Scientific Chemicals

2-Iodobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A13558.18
50 g, Each
Quantity
Catalog number: A13558.18
also known as A13558-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
609-67-6
IUPAC Name
2-iodobenzoyl chloride
Molecular Formula
C7H4ClIO
InChI Key
MVIVDSWUOGNODP-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC=CC=C1I
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Melting Point (clear melt)
27.0-33.0?C
Appearance (Color)
White to cream
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or fused/lumpy solid

Description

2-Iodobenzoyl chloride has been used in palladium-catalyzed synthesis of carbomethoxy functional group-induced isoindolin-1-ones, preparation of starting materials required for the synthesis of novel cyclic derivatives of pentavalent iodine, benziodazole oxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodobenzoyl chloride has been used in palladium-catalyzed synthesis of carbomethoxy functional group-induced isoindolin-1-ones, preparation of starting materials required for the synthesis of novel cyclic derivatives of pentavalent iodine, benziodazole oxides.

Solubility
Reacts with water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text