Thermo Scientific Chemicals

Silicon carbide powder, superfine, 600 grit, Thermo Scientific Chemicals

Catalog number: A13561.36
500 g, Each
Thermo Scientific Chemicals

Silicon carbide powder, superfine, 600 grit, Thermo Scientific Chemicals

Catalog number: A13561.36
500 g, Each
Quantity
Catalog number: A13561.36
also known as A13561-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
409-21-2
IUPAC Name
methanidylidynesilylium
Molecular Formula
CSi
InChI Key
HBMJWWWQQXIZIP-UHFFFAOYSA-N
SMILES
[C-]#[Si+]
Assay from Supplier's CofA
≥97.5% SiC
Impurity content
Fe₂O₃: ≤0.5%
Impurity content
Free C: ≤0.5%
Particle Size
Conforms to FEPA Micro grit standard
Particle Size
mean (d50) 9.3 ? 1 ?m

Description

Silicon carbide powder is used for its hardness in abrasive machining processes such as grinding, honing, water-jet cutting and sandblasting. And is also used as a support and shelving material in high temperature kilns such as for firing ceramics, glass fusing, or glass casting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silicon carbide powder is used for its hardness in abrasive machining processes such as grinding, honing, water-jet cutting and sandblasting. And is also used as a support and shelving material in high temperature kilns such as for firing ceramics, glass fusing, or glass casting.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text