Phenyl phosphorodiamidate, 98+%, Thermo Scientific Chemicals
Phenyl phosphorodiamidate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenyl phosphorodiamidate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number A13562.18
also known as A13562-18
Price (USD)/ Each
1,645.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
1,645.00
Add to cart
Phenyl phosphorodiamidate, 98+%, Thermo Scientific Chemicals
Catalog numberA13562.18
Price (USD)/ Each
1,645.00
-
Add to cart
Chemical Identifiers
CAS7450-69-3
IUPAC Name[(diaminophosphoryl)oxy]benzene
Molecular FormulaC6H9N2O2P
InChI KeyAYRRNFHDJUXLEQ-UHFFFAOYSA-N
SMILESNP(N)(=O)OC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (unspecified)≥96.0 to ≤104.0% (based on C, H, & N EA)
Elemental AnalysisC: 40.19-43.54% (theory 41.87%)
Elemental AnalysisH: 5.06-5.48% (theory 5.27%)
View more
Reagent for conversion of tautomeric keto-hydroxy groups (e.g. in N-heterocycles) directly to amino groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only