Ethylenediamine dihydrochloride, 98%, Thermo Scientific Chemicals
Ethylenediamine dihydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethylenediamine dihydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
100 g
500 g
Catalog number A13563.0E
also known as A13563-0E
Price (USD)/ Each
350.00
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
350.00
Add to cart
Ethylenediamine dihydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberA13563.0E
Price (USD)/ Each
350.00
-
Add to cart
Chemical Identifiers
CAS333-18-6
IUPAC Namedihydrogen ethane-1,2-diamine dichloride
Molecular FormulaC2H10Cl2N2
InChI KeyOHHBFEVZJLBKEH-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NCCN
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Identification (FTIR)Conforms
Appearance (Color)White
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)≤1%
Ethylenediamine dihydrochloride was used in modified ethylenediamine condensation method for fluorimetric determination of catecholamines. It was used to investigate the luminescence properties of complexes of EuIII and TbIII with ethylenediamine. It was used in the synthesis of 1,3,5-tris(4,5-dihydro-1H-imidazol-2-yl)benzene3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediamine dihydrochloride was used in modified ethylenediamine condensation method for fluorimetric determination of catecholamines. It was used to investigate the luminescence properties of complexes of EuIII and TbIII with ethylenediamine. It was used in the synthesis of 1,3,5-tris(4,5-dihydro-1H-imidazol-2-yl)benzene3

Solubility
Soluble in water 300 g/L (20°C).

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. M. Bounias. N-(1-Naphthyl)ethylenediamine dihydrochloride as a new reagent for nanomole quantification of sugars on thin-layer plates by a mathematical calibration process. Analytical Biochemistry. 1980, 102, (2), 291-295.
  2. Juana Bellanato. Infra-red spectra of ethylenediamine dihydrochloride and other amine hydrochlorides in alkali halide disks. Spectrochimica Acta. 1960, 16, (11-12), 1344-1357.