Thermo Scientific Chemicals

Ethylenediamine dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A13563.22
100 g, Each
Thermo Scientific Chemicals

Ethylenediamine dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A13563.22
100 g, Each
Quantity
Catalog number: A13563.22
also known as A13563-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
333-18-6
IUPAC Name
dihydrogen ethane-1,2-diamine dichloride
Molecular Formula
C2H10Cl2N2
InChI Key
OHHBFEVZJLBKEH-UHFFFAOYSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].NCCN
Form
Crystals or powder or crystalline powder or fused solid
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)
≤1%

Description

Ethylenediamine dihydrochloride was used in modified ethylenediamine condensation method for fluorimetric determination of catecholamines. It was used to investigate the luminescence properties of complexes of EuIII and TbIII with ethylenediamine. It was used in the synthesis of 1,3,5-tris(4,5-dihydro-1H-imidazol-2-yl)benzene3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediamine dihydrochloride was used in modified ethylenediamine condensation method for fluorimetric determination of catecholamines. It was used to investigate the luminescence properties of complexes of EuIII and TbIII with ethylenediamine. It was used in the synthesis of 1,3,5-tris(4,5-dihydro-1H-imidazol-2-yl)benzene3

Solubility
Soluble in water 300 g/L (20°C).

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text