Thermo Scientific Chemicals

4-(4-Nitrobenzyl)pyridine, 98+%, Thermo Scientific Chemicals

Catalog number: A13564.06
5 g, Each
Thermo Scientific Chemicals

4-(4-Nitrobenzyl)pyridine, 98+%, Thermo Scientific Chemicals

Catalog number: A13564.06
5 g, Each
Quantity
Catalog number: A13564.06
also known as A13564-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1083-48-3
IUPAC Name
4-[(4-nitrophenyl)methyl]pyridine
Molecular Formula
C12H10N2O2
InChI Key
MNHKUCBXXMFQDM-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(CC2=CC=NC=C2)C=C1
Appearance (Color)
Cream to pale yellow
Assay (GC)
≥98.0%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
67.0-74.0?C
Identification (FTIR)
Conforms

Description

4-(4-Nitrobenzyl)pyridine is used in spectroscopic method for the determination of phosgene. It is also used in tests for alkylating agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Nitrobenzyl)pyridine is used in spectroscopic method for the determination of phosgene. It is also used in tests for alkylating agents.

Solubility
Soluble in acetone. Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong bases and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text