2,4-Dihydroxypyridine, 97%, Thermo Scientific Chemicals
2,4-Dihydroxypyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Dihydroxypyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A13566.03
also known as A13566-03
Price (USD)/ Each
64.30
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
64.30
Add to cart
2,4-Dihydroxypyridine, 97%, Thermo Scientific Chemicals
Catalog numberA13566.03
Price (USD)/ Each
64.30
-
Add to cart
Chemical Identifiers
CAS626-03-9
IUPAC Name2-hydroxy-1,4-dihydropyridin-4-one
Molecular FormulaC5H5NO2
InChI KeyZEZJPIDPVXJEME-UHFFFAOYSA-N
SMILESOC1=CC(=O)C=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only