4-Methyl-5-thiazoleethanol, 98%, Thermo Scientific Chemicals
4-Methyl-5-thiazoleethanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methyl-5-thiazoleethanol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A13567.14
also known as A13567-14
Price (USD)/ Each
35.90
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
35.90
Add to cart
4-Methyl-5-thiazoleethanol, 98%, Thermo Scientific Chemicals
Catalog numberA13567.14
Price (USD)/ Each
35.90
-
Add to cart
Chemical Identifiers
CAS137-00-8
IUPAC Name2-(4-methyl-1,3-thiazol-5-yl)ethan-1-ol
Molecular FormulaC6H9NOS
InChI KeyBKAWJIRCKVUVED-UHFFFAOYSA-N
SMILESCC1=C(CCO)SC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid or viscous liquid
Appearance (Color)Clear colorless to yellow to orange to brown
Assay (GC)≥97.5%
CommentSpecification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)Conforms (non-U.S. specification)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only