Thermo Scientific Chemicals

(+)-Rutin trihydrate, 95%, Thermo Scientific Chemicals

Catalog number: A13570.36
500 g, Each
Thermo Scientific Chemicals

(+)-Rutin trihydrate, 95%, Thermo Scientific Chemicals

Catalog number: A13570.36
500 g, Each
Quantity
Catalog number: A13570.36
also known as A13570-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
250249-75-3
IUPAC Name
2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(3,4,5-trihydroxy-6-{[(3,4,5-trihydroxy-6-methyloxan-2-yl)oxy]methyl}oxan-2-yl)oxy]-4H-chromen-4-one trihydrate
Molecular Formula
C27H36O19
InChI Key
NLLBWFFSGHKUSY-UHFFFAOYNA-N
SMILES
O.O.O.CC1OC(OCC2OC(OC3=C(OC4=CC(O)=CC(O)=C4C3=O)C3=CC(O)=C(O)C=C3)C(O)C(O)C2O)C(O)C(O)C1O
Appearance (Color)
Yellow
Form
Powder
Assay (HPLC)
≥96.0%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
6.87-9.36% (2.5-3.5 waters)

Description

Rutin is the major mutagenic component of red wine, It is used in the activation of a mutagen by treatment both liver microsomal enzymes and fecal enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rutin is the major mutagenic component of red wine, It is used in the activation of a mutagen by treatment both liver microsomal enzymes and fecal enzymes.

Solubility
Soluble in DMSO, acetone, dichloromethane or ethyl acetate. Insoluble in water.

Notes
Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text