Thermo Scientific Chemicals

Rhodamine B

Catalog number: A13572.30
250 g, Each
Thermo Scientific Chemicals

Rhodamine B

Catalog number: A13572.30
250 g, Each
Quantity
Catalog number: A13572.30
also known as A13572-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
81-88-9
IUPAC Name
9-(2-carboxyphenyl)-3,6-bis(diethylamino)-10λ⁴-xanthen-10-ylium chloride
Molecular Formula
C28H31ClN2O3
InChI Key
PYWVYCXTNDRMGF-UHFFFAOYSA-N
SMILES
[Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC
Loss on Drying
≤10.0% (105?C)
lambda-max (UV-Vis)
545-560 nm (50:50 IMS:water)
Specific Absorbance (E 1%/1cm)
≥1700 (at lambda max, 50:50 IMS:water, UK-sourced material)
Comment
Material sourced in UK and US
Appearance (Color)
Purple or dark green

Description

Suitable for fluorescent labellingRhodamine B is used as a laser dye, metal chelating agent and an additive to drugs and cosmetics. It also serves as a staining fluorescent dye, which is used as a colorant in inks, paper and plastics. Further, it is used to make pink fluorescent pigments. It is also employed as a biomarker in oral rabies vaccines for wildlife such as raccoons. In addition to this, it is a useful fluorochrome for histology, fluorescence resonance energy transfer (FRET) and mitochondrial probe.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Suitable for fluorescent labellingRhodamine B is used as a laser dye, metal chelating agent and an additive to drugs and cosmetics. It also serves as a staining fluorescent dye, which is used as a colorant in inks, paper and plastics. Further, it is used to make pink fluorescent pigments. It is also employed as a biomarker in oral rabies vaccines for wildlife such as raccoons. In addition to this, it is a useful fluorochrome for histology, fluorescence resonance energy transfer (FRET) and mitochondrial probe.

Solubility
Soluble in water, benzene and ethanol. Slightly soluble in acetone, hydrochloric acid, sodium hydroxide, methyl cellosolve and alcohol.

Notes
Incompatible with strong oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text