Thermo Scientific Chemicals

4-Aminophenol, 98%, Thermo Scientific Chemicals

Catalog number: A13581.18
50 g, Each
Thermo Scientific Chemicals

4-Aminophenol, 98%, Thermo Scientific Chemicals

Catalog number: A13581.18
50 g, Each
Quantity
Catalog number: A13581.18
also known as A13581-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-30-8
IUPAC Name
4-aminophenol
Molecular Formula
C6H7NO
InChI Key
PLIKAWJENQZMHA-UHFFFAOYSA-N
SMILES
NC1=CC=C(O)C=C1
Appearance (Color)
White to cream to pale brown or pink to purple
Form
Powder
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Comment
Material darkens on storage and may contain some darker crystals/particles without loss of quality
Identification (FTIR)
Conforms

Description

4-Aminophenol is used as a dye for textiles, hair, furs and feathers. It is also used as a photographic developer. It finds application as a chemical intermediate in pharmaceuticals and dyes. Further, it acts as a final intermediate in the preparation of paracetamol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminophenol is used as a dye for textiles, hair, furs and feathers. It is also used as a photographic developer. It finds application as a chemical intermediate in pharmaceuticals and dyes. Further, it acts as a final intermediate in the preparation of paracetamol.

Solubility
Soluble in acetonitrile, ethyl acetate, acetone, hot water, dimethylsulfoxide and alkalies. Slightly soluble in toluene, diethyl ether, ethanol, trifluoroacetic acid and cold water. Insoluble in benzene and chloroform.

Notes
Incompatible with acids, chloroformates and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text