Thermo Scientific Chemicals

myo-Inositol, 98+%

Catalog number: A13586.0E
2500 g, Each
Thermo Scientific Chemicals

myo-Inositol, 98+%

Catalog number: A13586.0E
2500 g, Each
Quantity
Catalog number: A13586.0E
also known as A13586-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
87-89-8
IUPAC Name
cyclohexane-1,2,3,4,5,6-hexol
Molecular Formula
C6H12O6
InChI Key
CDAISMWEOUEBRE-UHFFFAOYSA-N
SMILES
OC1C(O)C(O)C(O)C(O)C1O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (unspecified)
≥98.0% [UK sourced, assay is silylated GC, US sourced, assay from vendor's CofA]
Comment
Material sourced in UK or US
Identification (FTIR)
Conforms (UK Sourced Material)

Description

Myo-Inositol is a growth factor for animals and microorganisms. It is the most abundant form of polyols that serves as a structural element of secondary messengers in eukaryotic cells. It is used as an adulterant in many illegal drugs like cocaine and methamphetamine. Further, it is used as a stand-in for cocaine on television and film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Myo-Inositol is a growth factor for animals and microorganisms. It is the most abundant form of polyols that serves as a structural element of secondary messengers in eukaryotic cells. It is used as an adulterant in many illegal drugs like cocaine and methamphetamine. Further, it is used as a stand-in for cocaine on television and film.

Solubility
Soluble in water. Slightly soluble in ethanol, dimethyl sulfoxide and alcohol. Insoluble in ether.    

Notes
Incompatible with strong oxidizing agents.    
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text