Triethyl orthoformate, 98%, Thermo Scientific Chemicals
Triethyl orthoformate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethyl orthoformate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 mL
500 mL
2500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A13587.0D
also known as A13587-0D
Price (USD)/ Each
430.65
Online exclusive
478.00 
Save 47.35 (10%)
-
Add to cart
Quantity:
10,000 mL
Request bulk or custom format
Price (USD)/ Each
430.65
Online exclusive
478.00 
Save 47.35 (10%)
Add to cart
Triethyl orthoformate, 98%, Thermo Scientific Chemicals
Catalog numberA13587.0D
Price (USD)/ Each
430.65
Online exclusive
478.00 
Save 47.35 (10%)
-
Add to cart
Chemical Identifiers
CAS122-51-0
IUPAC Name(diethoxymethoxy)ethane
Molecular FormulaC7H16O3
InChI KeyGKASDNZWUGIAMG-UHFFFAOYSA-N
SMILESCCOC(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.3890-1.3930 @ 20?C
Appearance (Color)Clear colorless
Triethyl orthoformate is used in Bodroux-Chichibabin aldehyde synthesis to prepare an aldehyde with one carbon higher by reacting with Grignard reagent. It is also used in the electrophilic formylation of activated aromatic species such as phenol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl orthoformate is used in Bodroux-Chichibabin aldehyde synthesis to prepare an aldehyde with one carbon higher by reacting with Grignard reagent. It is also used in the electrophilic formylation of activated aromatic species such as phenol.

Solubility
Miscible with alcohols, acetone, ethers and chloroform. Slightly miscible with water.

Notes
Moisture sensitive. Incompatible with acids and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Acid-catalyzed reaction with an arylamine gives the N-ethoxymethylene derivative (imidic ester); see, e.g.: Org. Synth. Coll., 4, 464 (1963); reduction (NaBH4) gives the monomethyl amine: Synthesis, 55 (1974).
  2. Due to their intrinsic dehydrating property, orthoformates are convenient reagents for acetalization of carbonyl compounds, under acid catalysis, e.g. tosic acid: J. Am. Chem. Soc., 60, 1905 (1938); J. Org. Chem., 20, 1695 (1955), or acidic ion-exchange resin: Synthesis, 348 (1974). Selective catalysis with NBS has been found to give high yields of acetals under almost neutral conditions: Synlett, 1456 (1999).
  3. In the presence of BF3 etherate, is a source of the diethoxycarbenium ion: Synth. Commun., 19, 2307 (1989), a selective alkylating agent and a useful intermediate for the diethoxymethylenation of ketones, to give protected ß-ketoaldehydes: J. Org. Chem., 46, 2557 (1981). For a similar reaction with enamines to give the same products, see: Chem. Lett., 1307 (1982); Bull. Chem. Soc. Jpn., 57, 1876 (1984).
  4. Orthoformates have also found use in the electrophilic formylation of activated aromatic species, e.g.: phenols (in the presence of AlCl3): Chem. Ber., 96, 308 (1963); pyrrole derivatives (with TFA): Austral. J. Chem., 25, 1979 (1972).
  5. For further reactions of alkyl orthoformates, see Trimethyl orthoformate, A13760.
  6. Yar, M.; Shahzad, S.; Siddiqi, S. A.; Mahmood, N.; Rauf, A.; Anwar, M. S.; Chaudhry, A. A.; Rehman, I. Triethyl orthoformate mediated a novel crosslinking method for the preparation of hydrogels for tissue engineering applications: characterization and in vitro cytocompatibility analysis. Mater. Sci. Eng., C 2015, 56, 154-164.
  7. Szczepankiewicz, W.; Kuźnik, N. Synthesis of 3-arylquinazolin-4(3H)-imines from 2-amino-N'-arylbenzamidines and triethyl orthoformate. Tetrahedron Lett. 2015, 56 (10), 1198-1199.