4-(3-Phenylpropyl)pyridine is used in electrochemical processes for a novel system of microdroplets of PPP immobilized at graphite and in mesoporous ceramic electrodes, PPP has been used to investigate electrochemically driven mono-, di-, and tri-carboxylate anion transfer at the 4-(3-phenylpropyl)pyridine/aqueous electrolyte interface for a triple phase boundary system at graphite electrodes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(3-Phenylpropyl)pyridine is used in electrochemical processes for a novel system of microdroplets of PPP immobilized at graphite and in mesoporous ceramic electrodes, PPP has been used to investigate electrochemically driven mono-, di-, and tri-carboxylate anion transfer at the 4-(3-phenylpropyl)pyridine/aqueous electrolyte interface for a triple phase boundary system at graphite electrodes.
Solubility
Difficult to mix.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Chris H. Senanayake.; George B. Smith.; Kenneth M. Ryan.; Laura E. Fredenburgh.; Ji Liu.; F.Edward Roberts.; David L. Hughes.; Robert D. Larsen.; Thomas R. Verhoeven.; Paul J. Reider. The role of 4-(3-phenylpropyl)pyridine N-Oxide (P3NO) in the manganese-salen-catalyzed asymmetric epoxidation of indene. Tetrahedron Letters. 1996, 37, (19), 3271-3274.
Michael J. Bonne.; Christopher Reynolds.; Stuart Yates.; Galyna Shul.; Joanna Niedziolka.; Marcin Opallo.; Frank Marken. The electrochemical ion-transfer reactivity of porphyrinato metal complexes in 4-(3-phenylpropyl)pyridine | water systems. New J. Chem. 2006, 30, 327-334.