Thermo Scientific Chemicals

6-Bromoveratraldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A13591.06
5 g, Each
Thermo Scientific Chemicals

6-Bromoveratraldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A13591.06
5 g, Each
Quantity
Catalog number: A13591.06
also known as A13591-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5392-10-9
IUPAC Name
2-bromo-4,5-dimethoxybenzaldehyde
Molecular Formula
C9H9BrO3
InChI Key
UQQROBHFUDBOOK-UHFFFAOYSA-N
SMILES
COC1=CC(Br)=C(C=O)C=C1OC
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Free acid (titration)
≤1.0%
Melting Point (clear melt)
143.5-152.5?C

Description

6-Bromoveratraldehyde was used in the preparation of benzylchromenylamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromoveratraldehyde was used in the preparation of benzylchromenylamines.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text