6-Bromoveratraldehyde, 97%, Thermo Scientific Chemicals
6-Bromoveratraldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Bromoveratraldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number A13591.22
also known as A13591-22
Price (USD)/ Each
691.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
691.00
Add to cart
6-Bromoveratraldehyde, 97%, Thermo Scientific Chemicals
Catalog numberA13591.22
Price (USD)/ Each
691.00
-
Add to cart
Chemical Identifiers
CAS5392-10-9
IUPAC Name2-bromo-4,5-dimethoxybenzaldehyde
Molecular FormulaC9H9BrO3
InChI KeyUQQROBHFUDBOOK-UHFFFAOYSA-N
SMILESCOC1=CC(Br)=C(C=O)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Free acid (titration)≤1.0%
Melting Point (clear melt)143.5-152.5?C
6-Bromoveratraldehyde was used in the preparation of benzylchromenylamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromoveratraldehyde was used in the preparation of benzylchromenylamines.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. James L. Charlton.; M. M. Alauddin. Asymmetric lignan synthesis. Isolariciresinol dimethyl ether. J. Org. Chem. 1986, 51, (18), 3490-3493.
  2. John B. Data.; Raymond D. Bennett. Alauddin. Synthesis of 2,3,6,11-Tetramethoxy-5(12H)-naphthacenone as an Analogue of Tetracycline. J. Med. Chem. 1961, 4, (2), 327-333.