Thermo Scientific Chemicals

2-Methyl-1,4-naphthoquinone, 98%, Thermo Scientific Chemicals

Catalog number: A13593.30
250 g, Each
Thermo Scientific Chemicals

2-Methyl-1,4-naphthoquinone, 98%, Thermo Scientific Chemicals

Catalog number: A13593.30
250 g, Each
Quantity
Catalog number: A13593.30
also known as A13593-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58-27-5
IUPAC Name
2-methyl-1,4-dihydronaphthalene-1,4-dione
Molecular Formula
C11H8O2
InChI Key
MJVAVZPDRWSRRC-UHFFFAOYSA-N
SMILES
CC1=CC(=O)C2=CC=CC=C2C1=O
Form
Powder or lumps
Identification (FTIR)
Conforms
Melting Point (clear melt)
102.0-109.0?C
Appearance (Color)
Yellow to green
Assay (GC)
≥97.5%

Description

2-Methyl-1,4-naphthoquinone is a prothrombogenic compound and is used as a model quinone in cell culture investigations. 2-Methyl-1,4-naphthoquinone is a synthetic analogue of of 1,4-naphthoquinone with a methyl group in the 2-position. 2-Methyl-1,4-naphthoquinone can be used as an oxidative injury inducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-1,4-naphthoquinone is a prothrombogenic compound and is used as a model quinone in cell culture investigations. 2-Methyl-1,4-naphthoquinone is a synthetic analogue of of 1,4-naphthoquinone with a methyl group in the 2-position. 2-Methyl-1,4-naphthoquinone can be used as an oxidative injury inducing agent.

Solubility
Soluble in methanol

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text