3',4'-(Methylenedioxy)acetophenone, 98%, Thermo Scientific Chemicals
3',4'-(Methylenedioxy)acetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3',4'-(Methylenedioxy)acetophenone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A13597.09
also known as A13597-09
Price (USD)/ Each
41.90
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
41.90
Add to cart
3',4'-(Methylenedioxy)acetophenone, 98%, Thermo Scientific Chemicals
Catalog numberA13597.09
Price (USD)/ Each
41.90
-
Add to cart
Chemical Identifiers
CAS3162-29-6
IUPAC Name1-(2H-1,3-benzodioxol-5-yl)ethan-1-one
Molecular FormulaC9H8O3
InChI KeyBMHMKWXYXFBWMI-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C2OCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (GC)≥97.5%
FormPowder
Melting Point (clear melt)83.0-90.0?C
3?,4?-(Methylenedioxy)acetophenone is used in the synthesis of N-ethyl-1-(3,4-methylenedioxyphenyl)ethylamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′,4′-(Methylenedioxy)acetophenone is used in the synthesis of N-ethyl-1-(3,4-methylenedioxyphenyl)ethylamine.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only