Thermo Scientific Chemicals

2-Methoxy-4-methylphenol, 98+%, Thermo Scientific Chemicals

Catalog number: A13603.18
50 g, Each
Thermo Scientific Chemicals

2-Methoxy-4-methylphenol, 98+%, Thermo Scientific Chemicals

Catalog number: A13603.18
50 g, Each
Quantity
Catalog number: A13603.18
also known as A13603-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
93-51-6
IUPAC Name
2-methoxy-4-methylphenol
Molecular Formula
C8H10O2
InChI Key
PETRWTHZSKVLRE-UHFFFAOYSA-N
SMILES
COC1=CC(C)=CC=C1O
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Comment
May turn yellow on storage
Form
Liquid

Description

2-Methoxy-4-methylphenol is used in preparation of renewable bis(cyanate) esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxy-4-methylphenol is used in preparation of renewable bis(cyanate) esters.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text