Thermo Scientific Chemicals

2-Hydroxybenzonitrile, 98%, Thermo Scientific Chemicals

Catalog number: A13606.22
100 g, Each
Thermo Scientific Chemicals

2-Hydroxybenzonitrile, 98%, Thermo Scientific Chemicals

Catalog number: A13606.22
100 g, Each
Quantity
Catalog number: A13606.22
also known as A13606-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
611-20-1
IUPAC Name
2-hydroxybenzonitrile
Molecular Formula
C7H5NO
InChI Key
CHZCERSEMVWNHL-UHFFFAOYSA-N
SMILES
OC1=CC=CC=C1C#N
Appearance (Color)
White to brown
Form
crystalline powder
Assay (Aqueous acid-base Titration)
> 97.5%
Assay (GC)
> 97.5%
Solution Test
10% in methanol: clear

Description

2-Hydroxybenzonitrile is used as starting reagent during the synthesis of mono-alkoxyphenyloxazoline, furanoside.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxybenzonitrile is used as starting reagent during the synthesis of mono-alkoxyphenyloxazoline, furanoside.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text