Thermo Scientific Chemicals

2-Bromobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A13608.30
250 g, Each
Thermo Scientific Chemicals

2-Bromobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A13608.30
250 g, Each
Quantity
Catalog number: A13608.30
also known as A13608-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
88-65-3
IUPAC Name
2-bromobenzoic acid
Molecular Formula
C7H5BrO2
InChI Key
XRXMNWGCKISMOH-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC=C1Br
Form
Powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (Silylated GC)
≥97.5%
Melting Point (clear melt)
146.0-152.0?C
Appearance (Color)
White to cream

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text