Thermo Scientific Chemicals

2-Chlorobenzoylacetonitrile, 95%, Thermo Scientific Chemicals

Catalog number: A13611.22
100 g, Each
Thermo Scientific Chemicals

2-Chlorobenzoylacetonitrile, 95%, Thermo Scientific Chemicals

Catalog number: A13611.22
100 g, Each
Quantity
Catalog number: A13611.22
also known as A13611-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
40018-25-5
IUPAC Name
3-(2-chlorophenyl)-3-oxopropanenitrile
Molecular Formula
C9H6ClNO
InChI Key
SBSWHTFHLWSSQS-UHFFFAOYSA-N
SMILES
ClC1=CC=CC=C1C(=O)CC#N
Appearance (Color)
Cream to yellow to orange to pale brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text