Thermo Scientific Chemicals

3-Dimethylaminobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A13612.14
25 g, Each
Thermo Scientific Chemicals

3-Dimethylaminobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A13612.14
25 g, Each
Quantity
Catalog number: A13612.14
also known as A13612-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-64-9
IUPAC Name
3-(dimethylamino)benzoic acid
Molecular Formula
C9H11NO2
InChI Key
NEGFNJRAUMCZMY-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=CC(=C1)C(O)=O
Melting Point (clear melt)
147.5-153.5?C
Assay (Silylated GC)
≥97.5%
Appearance (Color)
Pale cream to cream to brown
Form
Powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

3-Dimethylaminobenzoic acid is used as a benzoic acid derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Dimethylaminobenzoic acid is used as a benzoic acid derivative.

Solubility
Soluble in methanol (50 mg/ml). Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to moisture. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text