Thermo Scientific Chemicals

Trifluoroacetic anhydride, 99+%, Thermo Scientific Chemicals

Catalog number: A13614.22
100 g, Each
Thermo Scientific Chemicals

Trifluoroacetic anhydride, 99+%, Thermo Scientific Chemicals

Catalog number: A13614.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A13614.22
also known as A13614-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
407-25-0
IUPAC Name
2,2,2-trifluoroacetyl 2,2,2-trifluoroacetate
Molecular Formula
C4F6O3
InChI Key
QAEDZJGFFMLHHQ-UHFFFAOYSA-N
SMILES
FC(F)(F)C(=O)OC(=O)C(F)(F)F
Form
Liquid
Chloride content
≤0.0050%
Impurity content
Sulphate, SO₄: ≤0.0500%
Assay from Supplier's CofA
≥99.0%
Impurity content
Fluoride, F: ≤0.0010%

Description

Trifluoroacetic anhydride is used for the introduction of trifluoroacetyl group in organic synthesis. It is involved in the preparation of N- and O-trifluoroacetyl derivatives of a wide range of biologically active compounds for gas chromatography analysis. It plays an important role as desiccant for trifluoroacetic acid. It is also used in the oxidation of aldehydes to acids, esters, amides as well as in the protection of alcohols and amines. In addition, it is used as analytical reagents, solvents and dehydration condensing agent. It serves as an intermediate of fluorine fine chemicals, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trifluoroacetic anhydride is used for the introduction of trifluoroacetyl group in organic synthesis. It is involved in the preparation of N- and O-trifluoroacetyl derivatives of a wide range of biologically active compounds for gas chromatography analysis. It plays an important role as desiccant for trifluoroacetic acid. It is also used in the oxidation of aldehydes to acids, esters, amides as well as in the protection of alcohols and amines. In addition, it is used as analytical reagents, solvents and dehydration condensing agent. It serves as an intermediate of fluorine fine chemicals, pharmaceuticals and agrochemicals.

Solubility
Miscible with benzene, dichloromethane, diethyl ether, dimethylformamide, terahydrofuran and acetonitrile.

Notes
Hygroscopic and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases, strong acids, and alcohols.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text