4'-tert-Butylacetophenone, 98%, Thermo Scientific Chemicals
4'-tert-Butylacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4'-tert-Butylacetophenone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number A13615.06
also known as A13615-06
Price (USD)/ Each
97.40
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
97.40
Add to cart
4'-tert-Butylacetophenone, 98%, Thermo Scientific Chemicals
Catalog numberA13615.06
Price (USD)/ Each
97.40
-
Add to cart
Chemical Identifiers
CAS943-27-1
IUPAC Name1-(4-tert-butylphenyl)ethan-1-one
Molecular FormulaC12H16O
InChI KeyUYFJYGWNYQCHOB-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C=C1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5185-1.5225 @ 20?C
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to pale yellow
4'-tert-Butylacetophenone is used in the synthesis of 2-pyridone derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-tert-Butylacetophenone is used in the synthesis of 2-pyridone derivatives.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Mats Larhed.; Anders Hallberg. Microwave-Promoted Palladium-Catalyzed Coupling Reactions. J. Org. Chem. 1996, 61 (26), 9582-9584.
  2. Alain Schlatter.; Wolf-D. Woggon. Enantioselective Transfer Hydrogenation of Aliphatic Ketones Catalyzed by Ruthenium Complexes Linked to the Secondary Face of β-Cyclodextrin. Advanced Synthesis & Catalysis. 2008, 350 (7-8), 995-1000.