L-Phenylalanine ethyl ester hydrochloride is used as an enzyme substrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Phenylalanine ethyl ester hydrochloride is used as an enzyme substrate.
Solubility
Soluble in methanol and water.
Notes
Keep container tightly sealed. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature. Protect from humidity and water.
RUO – Research Use Only
Martin J. O’Donnell.; Robin L. Polt. A mild and efficient route to Schiff base derivatives of amino acids. J. Org. Chem. 1982, 47 (132), 2663-2666.
Yasuo Ariyoshi.; Tetsuo Yamatani.; Noboru Uchiyama.; Yohko Adachi.; Naotake Sato. The Synthesis of a Sweet Peptide, α-L-Aspartyl-L-phenylalanine Methyl Ester, without the Use of Protecting Groups. Bulletin of the Chemical Society of Japan. 1973, 46 (6), 1893-1895.