Thermo Scientific Chemicals

L-Phenylalanine ethyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A13618.22
100 g, Each
Thermo Scientific Chemicals

L-Phenylalanine ethyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A13618.22
100 g, Each
Quantity
Catalog number: A13618.22
also known as A13618-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3182-93-2
IUPAC Name
hydrogen ethyl (2S)-2-amino-3-phenylpropanoate chloride
Molecular Formula
C11H16ClNO2
InChI Key
FPFQPLFYTKMCHN-PPHPATTJSA-N
SMILES
[H+].[Cl-].CCOC(=O)[C@@H](N)CC1=CC=CC=C1
Appearance (Color)
White to pale cream or pale blue
Form
Powder or crystalline powder or crystals or needles
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Assay (Freebase GC)
≥97.5%
Water Content (Karl Fischer Titration)
≤1%

Description

L-Phenylalanine ethyl ester hydrochloride is used as an enzyme substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Phenylalanine ethyl ester hydrochloride is used as an enzyme substrate.

Solubility
Soluble in methanol and water.

Notes
Keep container tightly sealed. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature. Protect from humidity and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text