2-Methoxybenzhydrazide, 98+%, Thermo Scientific Chemicals
2-Methoxybenzhydrazide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methoxybenzhydrazide, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number A13619.18
also known as A13619-18
Price (USD)/ Each
508.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
508.00
Add to cart
2-Methoxybenzhydrazide, 98+%, Thermo Scientific Chemicals
Catalog numberA13619.18
Price (USD)/ Each
508.00
-
Add to cart
Chemical Identifiers
CAS7466-54-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow or pale orange to brown
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
Assay (GC)≥98%
2-Methoxybenzhydrazide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxybenzhydrazide is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
RUO – Research Use Only

General References:

  1. Eric Therrien.; Guillaume Larouche.; Sukhdev Manku.; Martin Allan.; Natalie Nguyen.; Sylvia Styhler.; Marie-France Rober.; Anne-Christine Goulet.; Jeffrey M. Besterman.; Hannah Nguyen.; Amal Wahhab. 1,2-Diamines as inhibitors of co-activator associated arginine methyltransferase 1 (CARM1). Bioorganic & Medicinal Chemistry Letters. 2009, 19 (23), 6725-6732.
  2. Yuichi Kajiwara.; Atsushi Nagai.; Kazuo Tanaka.; Yoshiki Chujo. Efficient simultaneous emission from RGB-emitting organoboron dyes incorporated into organic-inorganic hybrids and preparation of white light-emitting materials. J. Mater. Chem. C. 2013, 1 (29), 4437-4444.