Thermo Scientific Chemicals

Flavone, 99%, Thermo Scientific Chemicals

Catalog number: A13627.03
1 g, Each
Thermo Scientific Chemicals

Flavone, 99%, Thermo Scientific Chemicals

Catalog number: A13627.03
1 g, Each
Quantity
Catalog number: A13627.03
also known as A13627-03
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
525-82-6
IUPAC Name
2-phenyl-4H-chromen-4-one
Molecular Formula
C15H10O2
InChI Key
VHBFFQKBGNRLFZ-UHFFFAOYSA-N
SMILES
O=C1C=C(OC2=CC=CC=C12)C1=CC=CC=C1
Form
Powder
Melting Point (clear melt)
95-98?C
Appearance (Color)
White to pale cream
Assay (GC)
≥98.5%

Description

Flavone is a potentially useful biochemical for cytochrome P450 studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Flavone is a potentially useful biochemical for cytochrome P450 studies.

Solubility
Soluble in acetone (25 mg/ml), methanol, alcohol, and chloroform. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers.Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text