3-Hydroxybenzoic acid is used in perfumery. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Hydroxybenzoic acid is used in perfumery. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
H. N. Shreenivasa Murthy.; B. K. Sadashiva. Ferroelectric and antiferroelectric switching behaviour in new unsymmetrical bent-core compounds derived from 3-hydroxybenzoic acid. J. Mater. Chem. 2005, 15 (20), 2056-2064.
L. Narváez.; E. Cano.; D. M. Bastidas. 3-Hydroxybenzoic acid as AISI 316L stainless steel corrosion inhibitor in a H2SO4-HF-H2O2 pickling solution. Journal of Applied Electrochemistry. 2005, 35 (5), 499-506.