Thermo Scientific Chemicals

alpha-Methylphenylacetonitrile, 96%, Thermo Scientific Chemicals

Catalog number: A13636.09
10 g, Each
Thermo Scientific Chemicals

alpha-Methylphenylacetonitrile, 96%, Thermo Scientific Chemicals

Catalog number: A13636.09
10 g, Each
Quantity
Catalog number: A13636.09
also known as A13636-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1823-91-2
IUPAC Name
2-phenylpropanenitrile
Molecular Formula
C9H9N
InChI Key
NVAOLENBKNECGF-UHFFFAOYNA-N
SMILES
CC(C#N)C1=CC=CC=C1
Refractive Index
1.5090-1.5150 @ 20?C
Assay (GC)
≥95.0%
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to orange

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text