Thermo Scientific Chemicals

Diethylamine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A13637.30
250 g, Each
Thermo Scientific Chemicals

Diethylamine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A13637.30
250 g, Each
Quantity
Catalog number: A13637.30
Price (USD)
Have Questions?

Chemical Identifiers

CAS
660-68-4
IUPAC Name
diethylazanium chloride
Molecular Formula
C4H12ClN
InChI Key
HDITUCONWLWUJR-UHFFFAOYSA-N
SMILES
[Cl-].CC[NH2+]CC
Assay (Titration ex Chloride)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms (non-U.S. specification)
Appearance (Color)
White to cream to pale brown
Form
Crystals or powder or crystalline powder or lumps
Assay (Titration ex Chloride)
≥98.5 to ≤101.5%

Description

Diethylamine hydrochloride acts as a precursor of atrazine and lysergic acid diethylamide. It is also used as a reactant in the production of dyes and pharmaceutical compounds such as ranitidine. Further, it is used in Mannich reaction with paraformaldehyde. In addition to this, it is employed in the synthesis of diethylaminoethyl (DEAE) cottons by reacting with cotton cellulose in the presence of sodium hydroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylamine hydrochloride acts as a precursor of atrazine and lysergic acid diethylamide. It is also used as a reactant in the production of dyes and pharmaceutical compounds such as ranitidine. Further, it is used in Mannich reaction with paraformaldehyde. In addition to this, it is employed in the synthesis of diethylaminoethyl (DEAE) cottons by reacting with cotton cellulose in the presence of sodium hydroxide.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text