Thermo Scientific Chemicals

2-(4-Chlorophenyl)indole, 98%, Thermo Scientific Chemicals

Catalog number: A13638.03
1 g, Each
Thermo Scientific Chemicals

2-(4-Chlorophenyl)indole, 98%, Thermo Scientific Chemicals

Catalog number: A13638.03
1 g, Each
Quantity
Catalog number: A13638.03
also known as A13638-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1211-35-4
IUPAC Name
2-(4-chlorophenyl)-1H-indole
Molecular Formula
C14H10ClN
InChI Key
KDNXKQSAAZNUCK-UHFFFAOYSA-N
SMILES
ClC1=CC=C(C=C1)C1=CC2=CC=CC=C2N1
Assay (GC)
≥97.5%
Appearance (Color)
Cream
Melting Point (clear melt)
201.5-207.5?C
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text