Thermo Scientific Chemicals

5-Bromoisatin, 90+%, Thermo Scientific Chemicals

Catalog number: A13641.36
500 g, Each
Thermo Scientific Chemicals

5-Bromoisatin, 90+%, Thermo Scientific Chemicals

Catalog number: A13641.36
500 g, Each
Quantity
Catalog number: A13641.36
also known as A13641-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
87-48-9
IUPAC Name
5-bromo-2,3-dihydro-1H-indole-2,3-dione
Molecular Formula
C8H4BrNO2
InChI Key
MBVCESWADCIXJN-UHFFFAOYSA-N
SMILES
BrC1=CC=C2NC(=O)C(=O)C2=C1
Appearance (Color)
Orange
Assay (HPLC)
≥90.0%
Water Content (Karl Fischer Titration)
≤5%
Form
Powder
Identification (FTIR)
Conforms

Description

5-Bromoisatin is used in the synthesis of N-derivatives of 5-bromoisatin, N-substituted pyrroles, linear polyaryleneoxindoles, 5-bromodioxindole, cinchoninic acid derivatives, 3-hydroxyoxindole, S-benzyldithiocarbazate Schiff Bases, 5-bromooxindole and Morita-Baylis-Hillman adducts of isatin derivatives. It is an indole derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromoisatin is used in the synthesis of N-derivatives of 5-bromoisatin, N-substituted pyrroles, linear polyaryleneoxindoles, 5-bromodioxindole, cinchoninic acid derivatives, 3-hydroxyoxindole, S-benzyldithiocarbazate Schiff Bases, 5-bromooxindole and Morita-Baylis-Hillman adducts of isatin derivatives. It is an indole derivative.

Solubility
Soluble in N,N dimethylformamide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text