Thermo Scientific Chemicals

Chlorotrimethylsilane, 98%, Thermo Scientific Chemicals

Catalog number: A13651.AE
100 mL, Each
Thermo Scientific Chemicals

Chlorotrimethylsilane, 98%, Thermo Scientific Chemicals

Catalog number: A13651.AE
100 mL, Each
Quantity
Catalog number: A13651.AE
also known as A13651-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-77-4
IUPAC Name
chlorotrimethylsilane
Molecular Formula
C3H9ClSi
InChI Key
IJOOHPMOJXWVHK-UHFFFAOYSA-N
SMILES
C[Si](C)(C)Cl
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.3865-1.3905 @ 20°C

Description

Chlorotrimethylsilane is used to silanize laboratory glassware, which makes the surfaces more lipophilic. It is used in the production of trimethylsilyl halides, pseudohalides and various organic silicon compounds. It is also used to produce hexamethyldisilane by reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorotrimethylsilane is used to silanize laboratory glassware, which makes the surfaces more lipophilic. It is used in the production of trimethylsilyl halides, pseudohalides and various organic silicon compounds. It is also used to produce hexamethyldisilane by reduction.

Solubility
Miscible with ether, benzene, diethylether and perchloroethylene.

Notes
Incompatible with water, strong acids, strong oxidizing agents, strong bases, alcohols, aldehydes, amines, ketones and esters.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text