Thermo Scientific Chemicals

Diethyl phenylthiomethylphosphonate, 96%, Thermo Scientific Chemicals

Catalog number: A13660.18
50 g, Each
Thermo Scientific Chemicals

Diethyl phenylthiomethylphosphonate, 96%, Thermo Scientific Chemicals

Catalog number: A13660.18
50 g, Each
Quantity
Catalog number: A13660.18
also known as A13660-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
38066-16-9
IUPAC Name
diethyl [(phenylsulfanyl)methyl]phosphonate
Molecular Formula
C11H17O3PS
InChI Key
FBUXEPJJFVDUFE-UHFFFAOYSA-N
SMILES
CCOP(=O)(CSC1=CC=CC=C1)OCC
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Refractive Index
1.5310-1.5370 @ 20?C
Assay (GC)
≥95.0%

Description

Diethyl phenylthiomethylphosphonate is used to produce Diethylphosphorylmethylphenylsulfoxid, Chemical reagents, pharmaceutical research, pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl phenylthiomethylphosphonate is used to produce Diethylphosphorylmethylphenylsulfoxid, Chemical reagents, pharmaceutical research, pharmaceutical intermediates.

Solubility
Difficult to mix.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text