Thermo Scientific Chemicals

Iodoform, 99%, Thermo Scientific Chemicals

Catalog number: A13664.18
50 g, Each
Thermo Scientific Chemicals

Iodoform, 99%, Thermo Scientific Chemicals

Catalog number: A13664.18
50 g, Each
Quantity
Catalog number: A13664.18
also known as A13664-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-47-8
IUPAC Name
triiodomethane
Molecular Formula
CHI3
InChI Key
OKJPEAGHQZHRQV-UHFFFAOYSA-N
SMILES
IC(I)I
Appearance (Color)
Yellow
Form
Crystals or powder or crystalline powder
Assay (Titration ex Iodide)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms
Loss on Drying
≤0.5% (70?C/constant weight)

Description

Iodoform is use as a disinfectant. It is an active ingredient of ear powder along with other constituents like zinc oxide and propanoic acid, which prevent infection and facilitate the removal of ear hair in dogs and cats. In analytical chemistry, it is involved in the determination of iodoform in drinking water as well as to remove heavy metals like mercury from fluids. It plays an important role in the fermentation of bio mass. It is also used in root canal therapy for infected primary teeth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodoform is use as a disinfectant. It is an active ingredient of ear powder along with other constituents like zinc oxide and propanoic acid, which prevent infection and facilitate the removal of ear hair in dogs and cats. In analytical chemistry, it is involved in the determination of iodoform in drinking water as well as to remove heavy metals like mercury from fluids. It plays an important role in the fermentation of bio mass. It is also used in root canal therapy for infected primary teeth.

Solubility
Soluble in water, ether, acetic acid, ethanol, chloroform, ether, glycerol, carbon disulfide, olive oil, benzene and acetone. Slightly soluble in petroleum ether.

Notes
Light sensitive. Incompatible with strong bases, alkali metals, trong oxidizing agents and magnesium.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text