3-Bromoaniline, 98%, Thermo Scientific Chemicals
3-Bromoaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromoaniline, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number A13665.22
also known as A13665-22
Price (USD)/ Each
174.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
174.00
Add to cart
3-Bromoaniline, 98%, Thermo Scientific Chemicals
Catalog numberA13665.22
Price (USD)/ Each
174.00
-
Add to cart
Chemical Identifiers
CAS591-19-5
IUPAC Name3-bromoaniline
Molecular FormulaC6H6BrN
InChI KeyDHYHYLGCQVVLOQ-UHFFFAOYSA-N
SMILESNC1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.6230-1.6280 @ 20?C
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to yellow to orange to brown
FormLiquid
3-Bromoaniline is used in the synthesis of amino substituted quinazoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromoaniline is used in the synthesis of amino substituted quinazoline.

Solubility
Soluble in water (0.97 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Nielsen, S. F.; Peters, D.; Axelsson, O. The Suzuki Reaction Under Solvent-Free Conditions. Synthetic Communications: An International Journal for Rapid Communication of Synthetic Organic Chemistry. 2000, 30 (19), 3501-3509.
  2. Felix E. Goodson.; John F. Hartwig. Regiodefined Poly(N-arylaniline)s and Donor-Acceptor Copolymers via Palladium-Mediated Amination Chemistry. Macromolecules. 1998, 31 (5), 1700-1703.