4-Methyl-2-nitroaniline is used as a red azo dye.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methyl-2-nitroaniline is used as a red azo dye.
Solubility
Soluble in water (0.2 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
Michael P. Doyle.; Bernard Siegfried.; Joseph F. DellariaJr. Alkyl nitrite-metal halide deamination reactions. 2. Substitutive deamination of arylamines by alkyl nitrites and copper(II) halides. A direct and remarkably efficient conversion of arylamines to aryl halides. J. Org. Chem. 1977, 42 (14), 2426-2431.
Shoji Kajigaeshi.; Takaaki Kakinami.; Kazuhisa Inoue.; Manabu Kondo.; Hiroko Nakamura.; Masahiro Fujikawa.; Tsuyoshi Okamoto. Halogenation Using Quaternary Ammonium Polyhalides. VI. Bromination of Aromatic Amines by Use of Benzyltrimethylammonium Tribromide. Bulletin of the Chemical Society of Japan. 1988, 61 (2), 597-599.