Thermo Scientific Chemicals

m-Tolylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A13667.06
5 g, Each
Thermo Scientific Chemicals

m-Tolylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A13667.06
5 g, Each
Quantity
Catalog number: A13667.06
also known as A13667-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
637-04-7
IUPAC Name
hydrogen (3-methylphenyl)hydrazine chloride
Molecular Formula
C7H11ClN2
InChI Key
SHTBTGXNVWYODU-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CC1=CC=CC(NN)=C1
Appearance (Color)
Pale cream to cream to pale brown
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Form
Powder or fused solid
Assay (Freebase GC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text