Thermo Scientific Chemicals

L-(+)-Tartaric acid, 99%, Thermo Scientific Chemicals

Catalog number: A13668.22
100 g, Each
Thermo Scientific Chemicals

L-(+)-Tartaric acid, 99%, Thermo Scientific Chemicals

Catalog number: A13668.22
100 g, Each
Quantity
Catalog number: A13668.22
also known as A13668-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
87-69-4
IUPAC Name
2,3-dihydroxybutanedioic acid
Molecular Formula
C4H6O6
InChI Key
FEWJPZIEWOKRBE-UHFFFAOYNA-N
SMILES
OC(C(O)C(O)=O)C(O)=O
Form
Crystals or crystalline powder or powder
Assay from Supplier's CofA
≥98.5% (U.S. sourced material)
Comment
Material Sourced in the U.S. and in other countries
Identification (FTIR)
Conforms
Melting Point
164-172°C (non-U.S. sourced material)

Description

L-(+)-Tartaric acid is widely utilized in pharmaceutical industries. It is used in soft drinks, confectionaries, food products, gelatin desserts and as a buffering agent. It forms a compound, TiCl2(O-i-Pr)2 with Diels-Alder catalyst and acta as a chelate agent in metal industries. Owing to its efficient chelating property towards metal ions, it is used in farming and metal industries for complexing micronutrients and for cleaning metal surfaces, respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(+)-Tartaric acid is widely utilized in pharmaceutical industries. It is used in soft drinks, confectionaries, food products, gelatin desserts and as a buffering agent. It forms a compound, TiCl2(O-i-Pr)2 with Diels-Alder catalyst and acta as a chelate agent in metal industries. Owing to its efficient chelating property towards metal ions, it is used in farming and metal industries for complexing micronutrients and for cleaning metal surfaces, respectively.

Solubility
Soluble in water, methanol and acetone.

Notes
Incompatible with oxidizing agents, bases and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text