2-n-Octylthiophene, 98%, Thermo Scientific Chemicals
2-n-Octylthiophene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-n-Octylthiophene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
50 g
Catalog number A13672.06
also known as A13672-06
Price (USD)/ Each
83.80
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
83.80
Add to cart
2-n-Octylthiophene, 98%, Thermo Scientific Chemicals
Catalog numberA13672.06
Price (USD)/ Each
83.80
-
Add to cart
Chemical Identifiers
CAS880-36-4
IUPAC Name2-octylthiophene
Molecular FormulaC12H20S
InChI KeyGIFWAJGKWIDXMY-UHFFFAOYSA-N
SMILESCCCCCCCCC1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4900-1.4940 @ 20*C
Assay (GC)≥97.5%
FormLiquid
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only