Thermo Scientific Chemicals

2,4-Difluoroaniline, 99%, Thermo Scientific Chemicals

Catalog number: A13673.22
100 g, Each
Thermo Scientific Chemicals

2,4-Difluoroaniline, 99%, Thermo Scientific Chemicals

Catalog number: A13673.22
100 g, Each
Quantity
Catalog number: A13673.22
also known as A13673-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
367-25-9
IUPAC Name
2,4-difluoroaniline
Molecular Formula
C6H5F2N
InChI Key
CEPCPXLLFXPZGW-UHFFFAOYSA-N
SMILES
NC1=CC=C(F)C=C1F
Appearance (Color)
Clear colorless to pale yellow to pale brown or pink or purple
Form
Liquid
Assay (GC)
≥98.5%
Refractive Index
1.5060-1.5090 @ 20°C

Description

2,4-Difluoroaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluoroaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text