Thermo Scientific Chemicals

Chlorotriphenylsilane, 96%, Thermo Scientific Chemicals

Catalog number: A13678.06
5 g, Each
Thermo Scientific Chemicals

Chlorotriphenylsilane, 96%, Thermo Scientific Chemicals

Catalog number: A13678.06
5 g, Each
Quantity
Catalog number: A13678.06
also known as A13678-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
76-86-8
IUPAC Name
chlorotriphenylsilane
Molecular Formula
C18H15ClSi
InChI Key
MNKYQPOFRKPUAE-UHFFFAOYSA-N
SMILES
Cl[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder or lumps or chunks or fused solid
Identification (FTIR)
Conforms
Assay (GC)
≥95.0%
Melting Point (clear melt)
87.0-99.0?C
Appearance (Color)
Colorless to white

Description

Chlorotriphenylsilane is used as a silylating agent. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorotriphenylsilane is used as a silylating agent. It is also used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store under dry inert gas. It is sensitive to moisture. Incompatible with bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text